ENAH5802DD05 Enamine

4-tert-butylpyridine-2-carbaldehyde

  • MDL number MFCD18254152
  • Cas Number : 67141-22-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2139574

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2139574
Availability details for
Quantity
  • Availability
  • Add to Cart