ENAH5802DD07 Enamine

3-cyclopropyl-1-ethyl-1H-pyrazole-5-carbaldehyde

  • MDL number MFCD32063790
  • Cas Number : 2230672-24-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7444805

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7444805
Availability details for
Quantity
  • Availability
  • Add to Cart