ENAH5802DD08 Enamine

5H,6H,7H,8H,9H-imidazo[1,2-a]azepine-2-carbaldehyde

  • MDL number MFCD18821630
  • Cas Number : 623564-91-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6512908

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6512908
Availability details for
Quantity
  • Availability
  • Add to Cart