ENAH5802DD18 Enamine

6-phenylhex-5-ynal

  • MDL number MFCD24673733
  • Cas Number : 130602-09-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7444373

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7444373
Availability details for
Quantity
  • Availability
  • Add to Cart