ENAH5802DD1B Enamine

1-phenyl-1H-1,2,4-triazole-5-carbaldehyde

  • MDL number MFCD29958237
  • Cas Number : 111340-51-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-179102

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-179102
Availability details for
Quantity
  • Availability
  • Add to Cart