ENAH5802DD1E Enamine

2-ethyl-1H-indole-3-carbaldehyde

  • MDL number MFCD00049344
  • Cas Number : 95202-42-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7444747

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7444747
Availability details for
Quantity
  • Availability
  • Add to Cart