ENAH5802DD21 Enamine

1-methyl-2-oxo-2,3-dihydro-1H-indole-5-carbaldehyde

  • MDL number MFCD19596950
  • Cas Number : 933728-55-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-193173

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-193173
Availability details for
Quantity
  • Availability
  • Add to Cart