ENAH5802DD23 Enamine

5-(5-methylfuran-2-yl)-1H-pyrazole-4-carbaldehyde

  • MDL number MFCD05181548
  • Cas Number : 1006464-09-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-25648

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-25648
Availability details for
Quantity
  • Availability
  • Add to Cart