ENAH5802DD26 Enamine

2,6-dimethoxy-3-methylbenzaldehyde

  • MDL number MFCD24677708
  • Cas Number : 251654-46-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7355664

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7355664
Availability details for
Quantity
  • Availability
  • Add to Cart