ENAH5802DD2C Enamine

1-(2,2-dimethylpropyl)piperidine-4-carbaldehyde

  • MDL number MFCD24686199
  • Cas Number : 917898-69-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2647082

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2647082
Availability details for
Quantity
  • Availability
  • Add to Cart