ENAH5802DD2D Enamine

2-fluoro-4,6-dimethoxybenzaldehyde

  • MDL number MFCD18906188
  • Cas Number : 214492-73-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 86-87
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6739566

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6739566
Availability details for
Quantity
  • Availability
  • Add to Cart