ENAH5802DD2E Enamine

3-formyl-1-methyl-1H-indole-2-carbonitrile

  • MDL number MFCD22063873
  • Cas Number : 154379-85-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-1179979

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1179979
Availability details for
Quantity
  • Availability
  • Add to Cart