ENAH5802DD33 Enamine

1-methyl-4-phenyl-1H-1,2,3-triazole-5-carbaldehyde

  • MDL number MFCD23161469
  • Cas Number : 1374672-79-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7443442

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7443442
Availability details for
Quantity
  • Availability
  • Add to Cart