ENAH5802DD34 Enamine

2-hydroxy-4-(1H-pyrazol-1-yl)benzaldehyde

  • MDL number MFCD32069116
  • Cas Number : 1780164-03-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7438257

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7438257
Availability details for
Quantity
  • Availability
  • Add to Cart