ENAH5802DD37 Enamine

(2S)-2-[(tert-butyldimethylsilyl)oxy]propanal

  • MDL number MFCD11501227
  • Cas Number : 87727-28-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-4299848

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-4299848
Availability details for
Quantity
  • Availability
  • Add to Cart