ENAH5802DD45 Enamine

6-chloro-1,2-benzothiazole-3-carbaldehyde

  • MDL number MFCD27921778
  • Cas Number : 1823920-56-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6738879

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6738879
Availability details for
Quantity
  • Availability
  • Add to Cart