ENAH5802DD49 Enamine

2-methoxy-6-(1H-pyrazol-1-yl)benzaldehyde

  • Cas Number : 2375268-96-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7444948

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7444948
Availability details for
Quantity
  • Availability
  • Add to Cart