ENAH5802DD52 Enamine

1-cycloheptylpiperidine-4-carbaldehyde

  • MDL number MFCD24331956
  • Cas Number : 1556302-09-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-5378241

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-5378241
Availability details for
Quantity
  • Availability
  • Add to Cart