ENAH5802DD55 Enamine

tert-butyl 2-formyl-1-methyl-1H-imidazole-5-carboxylate

  • Cas Number : 293733-40-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7431231

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7431231
Availability details for
Quantity
  • Availability
  • Add to Cart