ENAH5802DD57 Enamine

1-[2-(diethylamino)ethyl]piperidine-4-carbaldehyde

  • MDL number MFCD16836783
  • Cas Number : 1062587-10-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7437670

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7437670
Availability details for
Quantity
  • Availability
  • Add to Cart