ENAH5802DD58 Enamine

6-(trifluoromethyl)-1H-indole-4-carbaldehyde

  • MDL number MFCD22553191
  • Cas Number : 1100215-61-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-5408733

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-5408733
Availability details for
Quantity
  • Availability
  • Add to Cart