ENAH5802DD59 Enamine

tert-butyl 3-ethyl-3-formylazetidine-1-carboxylate

  • MDL number MFCD14582394
  • Cas Number : 1205750-55-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-4350234

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-4350234
Availability details for
Quantity
  • Availability
  • Add to Cart