ENAH5802DD5E Enamine

2-{1-[(tert-butyldimethylsilyl)oxy]cyclopropyl}acetaldehyde

  • MDL number MFCD30291549
  • Cas Number : 1253978-16-3
  • Molecular Weight : 214.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(1-((tert-butyldimethylsilyl)oxy)cyclopropyl)acetaldehyde
Inchi Code 1S/C11H22O2Si/c1-10(2,3)14(4,5)13-11(6-7-11)8-9-12/h9H,6-8H2,1-5H3
Inchi Key DHQHYAJJUXLKJM-UHFFFAOYSA-N

Description

Purity 95
Storage temp. -10
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7433507

Safety Information

signalword Warning
pcodes P210,P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P370+P378,P403+P233,P403+P235,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-7433507
pictograms GHS07
hstatements H227,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart