ENAH5802DD60 Enamine

3-(3-fluorophenoxy)benzaldehyde

  • MDL number MFCD19613813
  • Cas Number : 65295-60-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7431706

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7431706
Availability details for
Quantity
  • Availability
  • Add to Cart