ENAH5802DD61 Enamine

6,7-dimethoxynaphthalene-2-carbaldehyde

  • MDL number MFCD32066634
  • Cas Number : 66920-80-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7428142

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7428142
Availability details for
Quantity
  • Availability
  • Add to Cart