ENAH5802DD66 Enamine

ethyl 3-formylpyrazolo[1,5-a]pyridine-2-carboxylate

  • MDL number MFCD16652606
  • Cas Number : 151831-22-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7443211

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7443211
Availability details for
Quantity
  • Availability
  • Add to Cart