ENAH5802DD6C Enamine

methyl 3-formyl-1-benzothiophene-5-carboxylate

  • MDL number MFCD22560593
  • Cas Number : 530144-81-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-716446

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-716446
Availability details for
Quantity
  • Availability
  • Add to Cart