ENAH5802DD6D Enamine

3-(cyclohexylmethoxy)pyridazine-4-carbaldehyde

  • MDL number MFCD32066208
  • Cas Number : 2361634-09-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7433574

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7433574
Availability details for
Quantity
  • Availability
  • Add to Cart