ENAH5802DD6F Enamine

methyl 4-(3-formylphenoxy)butanoate

  • MDL number MFCD12168351
  • Cas Number : 143816-77-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7438427

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7438427
Availability details for
Quantity
  • Availability
  • Add to Cart