ENAH5802DE29 Enamine

1-[(E)-2-nitroethenyl]naphthalene

  • Cas Number : 37630-26-5
  • Molecular Weight : 199.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7440594

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7440594
Availability details for
Quantity
  • Availability
  • Add to Cart