ENAH5802DE38 Enamine

3-chloro-3-(4-nitrophenyl)prop-2-enenitrile

  • Cas Number : 126417-79-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-06215

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-06215
Availability details for
Quantity
  • Availability
  • Add to Cart