ENAH5802DE41 Enamine

4-(ethoxymethylidene)-1,2,3,4-tetrahydroisoquinoline-1,3-dione

  • Cas Number : 18630-93-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-15408

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-15408
Availability details for
Quantity
  • Availability
  • Add to Cart