ENAH5802DE52 Enamine

propan-2-yl 1-bromoethene-1-sulfonate

  • MDL number MFCD31924111
  • Cas Number : 2344679-21-4
  • Molecular Weight : 229.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6491113

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6491113
Availability details for
Quantity
  • Availability
  • Add to Cart