ENAH5802DE61 Enamine

1'-ethyl-6'-oxo-1',2',5',6'-tetrahydrospiro[cyclopentane-1,4'-pyrazolo[3,4-b]pyridine]-5'-carbonitrile

  • MDL number MFCD23146197
  • Cas Number : 1955554-20-7
  • Molecular Weight : 244.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-25476

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-25476
Availability details for
Quantity
  • Availability
  • Add to Cart