ENAH5802DE6D Enamine

ethyl 3-[(2-chlorophenyl)amino]-2-cyanoprop-2-enoate

  • Cas Number : 7510-65-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-40516

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-40516
Availability details for
Quantity
  • Availability
  • Add to Cart