ENAH5802DE74 Enamine

5-[(2-amino-1,3-thiazol-4-yl)methyl]-4-(prop-2-en-1-yl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD26143610
  • Cas Number : 690643-10-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-30401

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-30401
Availability details for
Quantity
  • Availability
  • Add to Cart