ENAH5802DE79 Enamine

6'-oxo-1'-(propan-2-yl)-1',2',5',6'-tetrahydrospiro[cyclopentane-1,4'-pyrazolo[3,4-b]pyridine]-5'-carbonitrile

  • MDL number MFCD23146196
  • Cas Number : 1955540-48-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-25475

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-25475
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart