ENAH5802DE98 Enamine

4-(ethoxymethylidene)-2-(pyridin-2-yl)-1,2,3,4-tetrahydroisoquinoline-1,3-dione

  • MDL number MFCD07366298
  • Cas Number : 750601-20-8
  • Molecular Weight : 294.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-04011

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-04011
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart