ENAH5802DEAA Enamine

1-benzyl-4-[(4-methoxyphenyl)methylidene]-2-sulfanyl-4,5-dihydro-1H-imidazol-5-one

  • Cas Number : 875424-21-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 16-17
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-14187

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-14187
Availability details for
Quantity
  • Availability
  • Add to Cart