ENAH5802DED1 Enamine

5-[(3-bromophenyl)methylidene]-3-(2,4-dichlorobenzoyl)-2-sulfanylideneimidazolidin-4-one

  • Cas Number : 565170-80-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-05629

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05629
Availability details for
Quantity
  • Availability
  • Add to Cart