ENAH5802DEE2 Enamine

(2E)-4-chlorobut-2-en-1-yl acetate

  • Cas Number : 34414-28-3
  • Molecular Weight : 148.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7436885

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7436885
Availability details for
Quantity
  • Availability
  • Add to Cart