ENAH5802DF4A Enamine

N-(2-butyl-2,3-dihydro-1H-pyrazol-3-ylidene)-2-chloroacetamide

  • Cas Number : 1379396-08-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-46834

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-46834
Availability details for
Quantity
  • Availability
  • Add to Cart