ENAH5802DF4F Enamine

3-(bromomethyl)-5-(2-methylpropyl)-1,2-oxazole

  • MDL number MFCD31924568
  • Cas Number : 2344685-12-5
  • Molecular Weight : 218.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6526719

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6526719
Availability details for
Quantity
  • Availability
  • Add to Cart