ENAH5802DF7F Enamine

methyl 2-[3-(3-chloropropyl)-4-oxo-1,3-thiazolidin-2-ylidene]acetate

  • Cas Number : 1235441-77-6
  • Molecular Weight : 249.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-57146

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-57146
Availability details for
Quantity
  • Availability
  • Add to Cart