ENAH5802DF83 Enamine

2-chloro-N-[3-(propylsulfanyl)-2,5-dihydro-1,2,4-thiadiazol-5-ylidene]acetamide

  • Cas Number : 302784-83-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-24771

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-24771
Availability details for
Quantity
  • Availability
  • Add to Cart