ENAH5802DFA4 Enamine

2-chloro-N-(2-{[4-(propan-2-yl)phenyl]methyl}-2,3-dihydro-1H-pyrazol-3-ylidene)acetamide

  • Cas Number : 1955564-27-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-46365

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-46365
Availability details for
Quantity
  • Availability
  • Add to Cart