ENAH5802DFBD Enamine

1,4-dibromo-2-(bromomethyl)benzene

  • MDL number MFCD11052870
  • Cas Number : 136105-40-3
  • Molecular Weight : 328.83

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 93-94
Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-120083

Safety Information

signalword Danger
pcodes P260,P264,P280,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P363,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-120083
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart