ENAH5802DFC1 Enamine

N-[3-(chloromethyl)-4-methyl-4,5-dihydro-1,2,4-thiadiazol-5-ylidene]-N'-phenylbenzenecarboximidamide

  • Cas Number : 925549-73-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-24254

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-24254
Availability details for
Quantity
  • Availability
  • Add to Cart