ENAH5802DFE7 Enamine

4-amino-3-ethyl-1H-pyrazole-5-carboxamide

  • MDL number MFCD16249838
  • Cas Number : 215298-74-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-87265

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-87265
Availability details for
Quantity
  • Availability
  • Add to Cart