ENAH5802E004 Enamine

5-(furan-2-yl)-1,2-oxazole-3-carboxamide

  • MDL number MFCD11643941
  • Cas Number : 1098361-48-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-194258

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-194258
Availability details for
Quantity
  • Availability
  • Add to Cart